Benzo[h]quinoline 97%

Warning

Synonyms: 7,8-Benzoquinolinealpha-Naphthoquinoline4-Azaphenanthrene

AC15407-0010
200010-832EA 33.9 USD
200010-832
Benzo[h]quinoline 97%
Benzo[h]quinoline

Appearance: White to brown Crystalline powder or crystals and/or chunks

Formula: C₁₃H₉N
MW: 179.22 g/mol
Boiling Pt: 338 °C (719 mmHg)
Melting Pt: 48…52 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004984
CAS Number: 230-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR