Quinazoline 98%
AC15398-0050
AC15398-0010
200032-734EA
44.4
USD
200032-734
200038-688
Quinazoline 98%
Quinazoline
Formula:
C₈H₆N₂ Melting Pt: 45…48 °C Flash Pt: 106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006712 CAS Number: 253-82-7 EINECS: 205-965-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...