6-Azauracil 99%

Danger

Synonyms: 1,2,4-Triazine-3,5(2H,4H)-dione

AC15397-0050
200010-782EA 69.2 USD
200010-782
6-Azauracil 99%
6-Azauracil
Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 276…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00006456
CAS Number: 461-89-2
EINECS: 207-318-0
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR