Ferrocenecarbaldehyde 98%

Danger

Synonyms: Ferrocenecarboxaldehyde

AC15353-0010
103525-822EA 24.7 USD
103525-822
Ferrocenecarbaldehyde 98%
Ferrocenecarbaldehyde
Formula: C₁₁H₁₀FeO
MW: 214.04 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00001429
EINECS: 235-158-1
UN: 3467
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR