Aminoacetaldehyde dimethyl acetal 99%

Danger

Synonyms: 2,2-Dimethoxyethanamine

AC15334-5000 AC15334-0250 AC15334-1000 AC15334-0025
103525-820EA 1945.38 USD
103525-820 200011-178 200005-726 200018-140
Aminoacetaldehyde dimethyl acetal 99%
Aminoacetaldehyde dimethyl acetal
Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 135…139 °C (95 mmHg)
Density: 0.97 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00008135
CAS Number: 22483-09-6
EINECS: 245-026-5
UN: 2733
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR