Rhodanine 99%

Warning

Synonyms: 2-Thioxothiazolidin-4-one2-Thioxo-1,3-thiazolidin-4-one2-Thioxo-4-thiazolidinone

AC15260-0250 AC15260-1000
200026-680EA 137 USD
200026-680 200014-340
Rhodanine 99%
Rhodanine
Formula: C₃H₃NOS₂
MW: 133.19 g/mol
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00005488
CAS Number: 141-84-4
EINECS: 205-505-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR