3-Aminocrotononitrile cis- and trans-mixture 96%

Warning

Synonyms: 3-Amino-2-butenenitrileDiacetonitrile3-Aminobut-2-enenitrile3-Iminobutyronitrile

AC15245-5000 AC15245-1000
200041-446EA 122 USD
200041-446 200023-158
3-Aminocrotononitrile cis- and trans-mixture 96%
3-Aminocrotononitrile (mixture of cis and trans isomers)
Formula: C₄H₆N₂
MW: 82.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00008071
CAS Number: 1118-61-2
EINECS: 214-266-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR