Diethyl vinylphosphonate 97%

AC15215-0100 AC15215-0250
200025-018EA 264 USD
200025-018 200025-016
Diethyl vinylphosphonate 97%
Diethyl vinylphosphonate
Formula: C₆H₁₃O₃P
MW: 164.14 g/mol
Boiling Pt: 202 °C (760 mmHg)
Density: 1.06 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
EINECS: 211-663-2
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR