4-Nitrophenethyl bromide 98%

Warning

Synonyms: 1-(2-Bromoethyl)-4-nitrobenzene2-(4-Nitrophenyl)-1-bromoethane

AC15162-0050 AC15162-0250
200017-672EA 41.4 USD
200017-672 200023-954
4-Nitrophenethyl bromide 98%
4-Nitrophenethyl bromide
Formula: C₈H₈BrNO₂
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00007386
CAS Number: 5339-26-4
EINECS: 226-271-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR