4-(4-Nitrophenylazo)resorcinol 90%

Warning

Synonyms: Azo VioletMagneson I4-((4-Nitrophenyl)diazenyl)benzene-1,3-diolAzoviolet

AC15151-1000 AC15151-0250
200029-474EA 133 USD
200029-474 200017-238
4-(4-Nitrophenylazo)resorcinol 90%
4-(4-Nitrophenylazo)resorcinol
Formula: C₁₂H₉N₃O₄
MW: 259.22 g/mol
Melting Pt: 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007310
CAS Number: 74-39-5
EINECS: 200-808-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR