Diethyl phenylmalonate 98%

Synonyms: Diethyl 2-phenylmalonate

AC15026-1000 AC15026-2500 AC15026-0010 AC15026-0025 AC15026-0050
200037-096EA 51.1 USD
200037-096 200025-006 103510-502 200049-014 200049-016
Diethyl phenylmalonate 98%
Diethyl phenylmalonate
Formula: C₁₃H₁₆O₄
MW: 236.27 g/mol
Boiling Pt: 170…172 °C (14 mmHg)
Melting Pt: 16…17 °C
Density: 1.09 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009144
CAS Number: 83-13-6
EINECS: 201-456-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR