Ethanolamine 99%

Danger

Synonyms: Monoethanolamine2-Aminoethanol

AC14958-0250 AC14958-0010 AC14958-0025
200001-134EA 57.4 USD
200001-134 200001-136 200019-032
Ethanolamine 99%
Ethanolamine
Formula: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: 10.3 °C
Density: 1.012 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
UN: 2491
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/11/67/2301167.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR