5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%

Warning

Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carboxaldehyde

AC14910-0250 AC14910-1000 AC14910-0050
103525-698EA 24.2 USD
103525-698 200036-296 200024-128
5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%
5-Norbornene-2-carboxaldehyde
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 67…70 °C (12 mmHg)
Density: 1.018 g/cm³
Flash Pt: 50.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00167568
CAS Number: 5453-80-5
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR