Aminoguanidinium hydrogen carbonate ≥98.5%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: 1-Aminoguanidinium hydrogen carbonateAminoguanidine hydrogen carbonate

AC14906-2500 AC14906-0010
200018-142EA 32.45 USD
200018-142 200030-404
Aminoguanidinium hydrogen carbonate ≥98.5%
Aminoguanidinium hydrogen carbonate
Formula: C₂H₈N₄O₃
MW: 136.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012949
CAS Number: 2582-30-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Residue after ignition ≤0.3 %
Titration with HClO4 ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR