3,3'-Dithiodipropionic acid 98%

Warning

Synonyms: 3,3-Dithiodipropionic Acid

AC14890-2500 AC14890-0500
200047-180EA 71.6 USD
200047-180 200022-994
3,3'-Dithiodipropionic acid 98%
3,3'-Dithiodipropionic acid
Formula: C₆H₁₀O₄S₂
MW: 210.28 g/mol
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00002780
CAS Number: 1119-62-6
EINECS: 214-284-0
UN: 3335
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR