Sodium mercaptoacetate (sodium thioglycolate) 98%

Danger

Synonyms: Thioglycolic AcidMercaptoacetic acid, sodium saltThioglycolic Acid Sodium Salt

AC14889-2500 AC14889-0500 AC14889-0010
103525-690EA 307 USD
103525-690 103525-692 200001-070
Sodium mercaptoacetate (sodium thioglycolate) 98%
Sodium mercaptoacetate (Sodium thioglycolate)
Formula: HSCH₂COONa
MW: 114.1 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043386
CAS Number: 367-51-1
EINECS: 206-696-4
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR