Methyl mercaptoacetate 95%

Danger

Synonyms: Methyl thioglycolate

AC14888-5000 AC14888-1000
103531-308EA 27.7 USD
103531-308 200007-014
Methyl mercaptoacetate 95%
Methyl mercaptoacetate
Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 156…159 °C (10 mmHg)
Melting Pt: –24 °C
Density: 1.187 g/cm³
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00004873
CAS Number: 2365-48-2
EINECS: 219-121-7
UN: 1228
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR