Diphenylphosphinic acid 99%

AC14871-0050 AC14871-0250
200018-870EA 142 USD
200018-870 200031-064
Diphenylphosphinic acid 99%
Diphenylphosphinic acid
Formula: C₁₂H₁₁O₂P
MW: 218.19 g/mol
Melting Pt: 193…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00002132
CAS Number: 1707-03-5
EINECS: 216-948-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR