Cyanoacetamide 99%

Warning

Synonyms: 2-Cyanoacetamide

AC14865-5000 AC14865-1000
200009-336EA 29 USD
200009-336 200009-338
Cyanoacetamide 99%
Cyanoacetamide
Formula: C₃H₄N₂O
MW: 84.08 g/mol
Melting Pt: 118…122 °C
Flash Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00008024
CAS Number: 107-91-5
EINECS: 203-531-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR