3-Amino-4-methoxyphenylsulfonyl fluoride ≥97%

Danger

Synonyms: 3-Amino-4-methoxybenzenesulfonyl fluoride3-Amino-4-methoxybenzene-1-sulfonyl fluoride3-Amino-4-methoxybenzenesulphonyl fluoride4-Methoxymetanilyl fluoride3-Amino-4-methoxybenzene-1-sulphonyl fluoride

AC14833-0250 AC14833-0100
200035-960EA 89.1 USD
200035-960 200023-890
3-Amino-4-methoxyphenylsulfonyl fluoride ≥97%
3-Amino-4-methoxyphenylsulphonyl fluoride
Formula: C₇H₈FNO₃S
MW: 205.2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007417
CAS Number: 498-74-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR