Nicotinic acid hydrazide 97%

Warning

Synonyms: (3-Pyridylcarbonyl)hydrazinePyridine-3-carbohydrazide

AC14820-0250 AC14820-1000
200020-076EA 39.1 USD
200020-076 200038-216
Nicotinic acid hydrazide 97%
Nicotinic acid hydrazide
Formula: C₆H₇N₃O
MW: 137.14 g/mol
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00006383
CAS Number: 553-53-7
EINECS: 209-041-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR