2,2-Bis(hydroxymethyl)propionic acid 98%

Warning

Synonyms: 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid

AC14797-0010 AC14797-0050 AC14797-5000
200004-846EA 43.8 USD
200004-846 103510-424 200028-082
2,2-Bis(hydroxymethyl)propionic acid 98%
2,2-Bis(hydroxymethyl)propionic acid
Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Melting Pt: 185…190 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004199
CAS Number: 4767-03-7
EINECS: 225-306-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR