Nitrilotriacetic acid trisodium salt monohydrate 99+%

Warning

Synonyms: Nitrilotriacetic acid, trisodium salt, monohydrate

AC14784-5000 AC14784-0025 AC14784-0250
200032-268EA 24.9 USD
200032-268 200020-090 103516-484
Nitrilotriacetic acid trisodium salt monohydrate 99+%
Nitrilotriacetic acid trisodium salt monohydrate
Formula: C₆H₆NNa₃O₆·1H₂O
MW: 275.1 g/mol
Melting Pt: >320 °C
Storage Temperature: Ambient
MDL Number: MFCD00150789
CAS Number: 18662-53-8
EINECS: 225-768-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR