2-(Bromoacetyl)naphthalene 98%

Danger

Synonyms: 2-Bromo-2'-acetonaphthoneα-Bromo-2'-acetonaphthone

AC14748-0100 AC14748-0500
200030-360EA 62.2 USD
200030-360 200018-076
2-(Bromoacetyl)naphthalene 98%
2-(Bromoacetyl)naphthalene
Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004109
CAS Number: 613-54-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR