Methyl-3-(methylthio)propionate 99+%

Synonyms: Methyl3-(methylthio)propionate

AC14700-0050
200031-850EA 15.3 USD
200031-850
Methyl-3-(methylthio)propionate 99+%
Methyl 3-(methylthio)propionate
Formula: C₅H₁₀O₂S
MW: 134.20 g/mol
Boiling Pt: 74…75 °C (13 mmHg)
Density: 1.07 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008565
EINECS: 236-883-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR