Cyclohexanemethylamine 97%
|
Danger
|
Synonyms:
(Aminomethyl)cyclohexane
, Cyclohexylmethanamine
, (Cyclohexylmethyl)amine
AC14657-0050
AC14657-0250
102774-198EA
59.1
USD
102774-198
102775-752
Cyclohexanemethylamine 97%
Cyclohexanemethylamine
Formula:
C₇H₁₅N MW: 113.2 g/mol Boiling Pt: 161…163 °C (1013 hPa) Melting Pt: –8 °C Density: 0.87 g/cm³ Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001520 CAS Number: 3218-02-8 UN: 2734 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...