Catechol violet, pure indicator grade

Synonyms: Pyrocatechol Violet

AC14654-0250 AC14654-0050
103467-404EA 213 USD
103467-404 200007-552
Catechol violet, pure indicator grade
Catechol violet
Formula: C₁₉H₁₄O₇S
Storage Temperature: Ambient
MDL Number: MFCD00005868
CAS Number: 115-41-3
EINECS: 204-088-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR