(±)-3-Aminoquinuclidine dihydrochloride 98%
|
Warning
|
Synonyms:
3-Aminoquinuclidine dihydrochloride
, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride
AC14615-0250
AC14615-0050
200035-254EA
146
USD
200035-254
200023-170
(±)-3-Aminoquinuclidine dihydrochloride 98%
(±)-3-Aminoquinuclidine dihydrochloride
Formula:
C₇H₁₆Cl₂N₂ MW: 199.1213 g/mol Melting Pt: 321…323 °C Storage Temperature: Ambient |
MDL Number:
MFCD00137395 CAS Number: 6530-09-2 EINECS: 229-424-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...