(1-Ethoxycarbonylethylidene)triphenylphosphorane ca. 94% balance TPPO

Danger

Synonyms: (Carbethoxyethylidene)triphenylphosphoraneEthyl 2-(triphenylphosphoranylidene)propanoate

AC14608-0050 AC14608-0250
103531-062EA 157 USD
103531-062 200008-188
(1-Ethoxycarbonylethylidene)triphenylphosphorane ca. 94% balance TPPO
(1-Ethoxycarbonylethylidene)triphenylphosphorane
Formula: C₂₃H₂₃O₂P
MW: 362.41 g/mol
Melting Pt: 157…159 °C
Storage Temperature: Freezer
MDL Number: MFCD00009160
CAS Number: 5717-37-3
EINECS: 227-215-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR