1-Vinyl-2-pyrrolidone 99% stabilized

Danger

Synonyms: N-Vinyl-2-pyrrolidinoneN-Vinyl-2-pyrrolidone

Stabilization: Stabilised with sodium hydroxide

AC14092-0100 AC14092-0025 AC14092-5000 AC14092-0050
103466-212EA 72.1 USD
103466-212 103466-210 103466-208 200058-146
1-Vinyl-2-pyrrolidone 99% stabilized
1-Vinyl-2-pyrrolidone
Formula: C₆H₉NO
MW: 111.1418 g/mol
Boiling Pt: 218 °C (760 mmHg)
Melting Pt: 13…14 °C
Density: 1.043 g/cm³
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00003197
CAS Number: 88-12-0
EINECS: 201-800-4
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR