Tropinone 99%

Danger

Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

AC14057-0500 AC14057-0100 AC14057-0050
200021-118EA 66 USD
200021-118 103531-052 103531-054
Tropinone 99%
Tropinone
Formula: C₈H₁₃NO
MW: 139.2 g/mol
Boiling Pt: 113 °C (25 mmHg)
Melting Pt: 41…44 °C
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005549
CAS Number: 532-24-1
EINECS: 208-530-6
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR