Trithiocyanuric acid 95%

Warning

AC14053-0050
200015-090EA 43.4 USD
200015-090
Trithiocyanuric acid 95%
Trithiocyanuric acid
Formula: C₃H₃N₃S₃
MW: 177.27 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006052
CAS Number: 638-16-4
EINECS: 211-322-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR