Triphenylmethyl mercaptan 97%

Warning

Synonyms: Triphenylmethanethiol

AC14041-0250 AC14041-5000 AC14041-1000
103516-468EA 1028 USD
103516-468 200007-996 200033-098
Triphenylmethyl mercaptan 97%
Triphenylmethyl mercaptan
Formula: C₁₉H₁₆S
MW: 276.40 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00004854
CAS Number: 3695-77-0
EINECS: 223-020-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR