Triethyl methanetricarboxylate 98%

AC13963-0250
200014-970EA 119 USD
200014-970
Triethyl methanetricarboxylate 98%
Triethyl methanetricarboxylate
Formula: C₁₀H₁₆O₆
MW: 232.23 g/mol
Boiling Pt: 253 °C (760 mmHg)
Melting Pt: 29 °C
Density: 1.1 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009150
CAS Number: 6279-86-3
EINECS: 228-477-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR