Triethyl-1,1,2-ethanetricarboxylate 99%

Synonyms: Triethyl ethane-1,1,2-tricarboxylate

AC13961-0050 AC13961-1000 AC13961-5000 AC13961-0250
103466-686EA 21.5 USD
103466-686 200000-942 200021-044 103466-688
Triethyl-1,1,2-ethanetricarboxylate 99%
Triethyl 1,1,2-ethanetricarboxylate
Formula: C₁₁H₁₈O₆
MW: 246.26 g/mol
Boiling Pt: 99 °C (.5 mmHg)
Density: 1.07 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009154
CAS Number: 7459-46-3
EINECS: 231-235-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR