Triethylene glycol dimethyl ether 99% stabilized

Danger

Synonyms: 2,5,8,11-Tetraoxadodecane

AC13960-0025 AC13960-0010 AC13960-2500
103466-696EA 466.15 USD
103466-696 200058-784 200058-782
Triethylene glycol dimethyl ether 99% stabilized
Triethylene glycol dimethyl ether
Formula: C₈H₁₈O₄
MW: 178.23 g/mol
Boiling Pt: 216 °C (760 mmHg)
Melting Pt: –45 °C
Density: 0.986 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008504
CAS Number: 112-49-2
EINECS: 203-977-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR