Tiglic acid 98%

Danger

Synonyms: (E)-2-Methylbut-2-enoic acidtrans-2,3-Dimethylacrylic acid

AC13897-1000 AC13897-0250
200032-988EA 33.7 USD
200032-988 200027-020
Tiglic acid 98%
Tiglic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Melting Pt: 61…65 °C
Density: 0.96 g/cm³
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066864
CAS Number: 80-59-1
EINECS: 201-295-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR