L-Threonine 98%

Synonyms: (2S,3R)-(-)-Threonine(2S,3R)-2-Amino-3-hydroxybutyric acidH-Thr-OH

AC13893-1000 AC13893-0050 AC13893-0250
103510-384EA 19.9 USD
103510-384 200000-918 200006-864
L-Threonine 98%
L-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR