Benzo[b]thiophene 97%

Warning

Synonyms: 1-BenzothiopheneThianaphthene

AC13862-0050 AC13862-1000 AC13862-0250
200014-798EA 89.6 USD
200014-798 200014-796 103525-516
Benzo[b]thiophene 97%
Benzo[b]thiophene
Formula: C₈H₆S
MW: 134.2 g/mol
Boiling Pt: 221…222 °C (760 mmHg)
Melting Pt: 28…32 °C
Density: 1.14 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005864
CAS Number: 95-15-8
EINECS: 202-395-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow to pink to orange Crystalline solid
Infrared spectrum Conforms
Melting point 28 to 33 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR