1-Benzyl-4-cyano-4-hydroxypiperidine 98%

AC13686-0100
200004-694EA 64.9 USD
200004-694
1-Benzyl-4-cyano-4-hydroxypiperidine 98%
1-Benzyl-4-cyano-4-hydroxypiperidine
Formula: C₁₃H₁₆N₂O
Melting Pt: 95…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00023752
CAS Number: 6094-60-6
EINECS: 228-043-2
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 95 to 102 °C
GC ≥97.5 %
Specific optical rotation -3° to -5° (20 °C, 589 nm) (c=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR