Sesamol-(3,4-(methylenedioxy)phenol) 98%

Synonyms: Benzo[d][1,3]dioxol-5-olSesamol3,4-Methylenedioxyphenol

AC13269-0050 AC13269-0250
200026-748EA 29.9 USD
200026-748 200014-406
Sesamol-(3,4-(methylenedioxy)phenol) 98%
Sesamol (3,4-(Methylenedioxy)phenol)
Formula: C₇H₆O₃
MW: 138.12 g/mol
Melting Pt: 63…65 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005827
CAS Number: 533-31-3
EINECS: 208-561-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder (May darken on storage)
Infrared spectrum Conforms
Melting point 63 to 65 °C
Water ≤0.2 % (K.F.)
HPLC ≥97.5 %
Specific optical rotation +80.0° to +90.0° (20 °C, 589 nm) (c=1, water, after 5 hours)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR