Rutin hydrate 97+%

AC13239-0050 AC13239-1000 AC13239-5000
200020-626EA 137 USD
200020-626 200050-792 200026-700
Rutin hydrate 97+%
Rutin hydrate
Formula: C₂₇H₃₀O₁₆
Melting Pt: 195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006830
CAS Number: 207671-50-9

Order Now

Specification Test Results

Appearance Yellow to yellow-green to straw yellow Powder
Infrared spectrum Conforms
Loss on drying 5.0 % to 9.0 % (500 mg, 105 °C, 3 h)
Ash ≤0.5 %
Titration with NaOH 97.0 to 102.0 % (On dry basis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR