Pyrrole-2-carboxylic acid 97%

Warning

AC13207-0250 AC13207-0050 AC13207-0010
102777-446EA 390 USD
102777-446 102775-750 102774-192
Pyrrole-2-carboxylic acid 97%
Pyrrole-2-carboxylic acid
Formula: C₅H₅NO₂
MW: 111.10 g/mol
Melting Pt: 207…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00005219
CAS Number: 634-97-9

Order Now

Specification Test Results

Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR