Nicotinaldehyde 98%

Danger

Synonyms: 3-Pyridinecarbaldehyde3-PyridinecarboxaldehydePyridine-3-carboxaldehyde

AC13183-5000 AC13183-1000 AC13183-0250
103525-434EA 360 USD
103525-434 200005-308 200000-822
Nicotinaldehyde 98%
Nicotinaldehyde
Formula: C₆H₅NO
MW: 107.11 g/mol
Boiling Pt: 95…97 °C (15 mmHg)
Density: 1.142 g/cm³
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006382
CAS Number: 500-22-1
EINECS: 207-900-4
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20 °C, 589 nm)
GC ≥97.5 %
pH 7.0 to 9.0
Titration with HClO4 94 to 106 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR