4-Picolyl chloride hydrochloride 97%

Danger

Synonyms: 4-(Chloromethyl)pyridine hydrochloride4-(Chloromethyl)-pyridinium chloride

AC13120-1000 AC13120-0100
200023-986EA 214 USD
200023-986 200010-624
4-Picolyl chloride hydrochloride 97%
4-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00012826
CAS Number: 1822-51-1
EINECS: 217-350-7
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %
GC ≥99.4 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR