Phthaloyl dichloride 90% remainder mainly phthalic acid

Danger

Synonyms: o-Phthaloyl dichloridePhthaloyl chloride

AC13111-0050
200044-328EA 19.5 USD
200044-328
Phthaloyl dichloride 90% remainder mainly phthalic acid
Phthaloyl dichloride
Formula: C₈H₄Cl₂O₂
MW: 203.02 g/mol
Boiling Pt: 269…271 °C (760 mmHg)
Melting Pt: 12 °C
Density: 1.4 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000666
CAS Number: 88-95-9
EINECS: 201-869-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5674 to 1.5694 (20 °C, 589 nm)
Titration RCOCl ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR