Phthalhydrazide 99%

Synonyms: 2,3-Dihydrophthalazine-1,4-dione

AC13106-1000 AC13106-0250
200063-376EA 32.2 USD
200063-376 200032-572
Phthalhydrazide 99%
Phthalhydrazide
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006888
CAS Number: 1445-69-8
EINECS: 215-893-4

Order Now

Specification Test Results

Appearance White Powder and/or chunks
Infrared spectrum Conforms
Loss on drying ≤1 %
Titration with NaOH 98.5 to 101.5 % (On dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR