Phthalazine 98%

Warning

Synonyms: Benzo[d]pyridazine

AC13105-0100
200014-036EA 115 USD
200014-036
Phthalazine 98%
Phthalazine
Formula: C₈H₆N₂
MW: 130.15 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006908
CAS Number: 253-52-1
EINECS: 205-963-2

Order Now

Specification Test Results

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 88 to 92 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR