Phenylphosphonic dichloride 97%

Danger

Synonyms: Dichlorophenylphosphine oxidePhenyldichlorophosphine oxide

AC13080-0010 AC13080-2500
103525-384EA 408 USD
103525-384 200007-344
Phenylphosphonic dichloride 97%
Phenylphosphonic dichloride
Formula: C₆H₅Cl₂OP
MW: 194.98 g/mol
Boiling Pt: 258 °C (760 mmHg)
Melting Pt: 3 °C
Density: 1.39 g/cm³
Flash Pt: 204 °C
Storage Temperature: Ambient
MDL Number: MFCD00002070
CAS Number: 824-72-6
EINECS: 212-534-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5570 to 1.5600 (20 °C, 589 nm)
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR