Phenylphosphonic acid 98%
|
Danger
|
Synonyms:
Phenylphosphonic Acid
AC13079-5000
AC13079-0025
AC13079-1000
200014-006EA
43.7
USD
200014-006
103525-382
200020-376
Phenylphosphonic acid 98%
Phenylphosphonic acid
Formula:
C₆H₇O₃P MW: 158.09 g/mol Melting Pt: 158…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002136 CAS Number: 1571-33-1 EINECS: 216-388-1 UN: 3261 ADR: 8,III |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 158 to 165 °C |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...